Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega cAMP-Dependent Protein Kinase, Catalytic Subunit
Click to view available options
Quantity:
2500 U
Description
- Purified Kinase for Protein Phosphorylation and Signal Transduction Studies
- Purified from recombinant E. coli expressing the catalytic subunit of bovine PKA; 90% pure
- Supplied at a concentration of 1.5-3mg/ml
Specifications
Specifications
Concentration | 1.5 to 3 mg/mL |
Color | Colorless |
Purity | 90% |
pH | 6.5 |
For Use With (Application) | Phosphorylate target Proteins or for in-vitro enzymological studies of neural and hormonal signal transduction |
Storage Buffer | 350mM Potassium Phosphate (pH 6.8), 0.1mM DTT |
Content And Storage | <-65°C |
Source | Recombinant E. coli strain |
Quantity | 2500 U |
Product Type | cAMP-dependent Protein Kinase |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction