Learn More
Promega Control Vectors for CheckMate™/Flexi™ Vector Mammalian Two-Hybrid Systems
Shop All Promega Corporation ProductsDescription
- Powerful System for Detecting Protein:Protein Interactions In Vivo
- Mammalian system for studying interactions in cell line of choice
- Study post-translational modifications including glycosylation, phosphorylation and acylation
Specifications
Specifications
| For Use With (Equipment) | CheckMate™/Flexi™ vector mammalian two-hybrid system |
| Storage Buffer | 10mM Tris HCl, 1mM EDTA (pH 7.4) |
| Content And Storage | -30°C to -10°C |
| Concentration | 1 μg/μL |
| Quantity | 1 Stab/ 20μg |
| Vector | pBIND, pACT neg ctrl |
| For Use With (Application) | Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.