Learn More
Description
Includes
DAB substrate and H2O2 plus plastic coverslips for easy sample handling
- A Modified TUNEL Assay for Detection of Apoptotic Cells In Situ
- Detects apoptosis in tissue sections or cell cultures
Specifications
Specifications
| Quantity | 20 Reactions |
| pH | 6.8 |
| Content And Storage | 1 x 20 each Plastic Coverslips, 1 x 4.8mL Equilibration Buffer, 1 x 20mL 20X SSC, 1 x 40μL Streptavidin HRP, 1 x 20μL Biotinylated Nucleotide Mix, 1 x 200μL DAB 20X Chromogen, 1 x 200μ. Compenents are stored at different temperatures |
| For Use With (Application) | Detection of apoptotic cells in situ at the single-cell level |
| Form | Fluid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.