Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega EGFR Kinase Enzyme System

Catalog No. p-4671644 Shop All Promega Corporation Products
Click to view available options
Quantity:
1/Ea.
10 μg

Includes

EGFR Kinase Enzyme System (V3831)

  • Kinase: EGFR, 10μg (Human, recombinant; amino acids 695–end). MW: ∽89kDa
  • Substrate: Poly (4:1 Glu, Tyr) Peptide
  • Others: Reaction Buffer, DTT, MnCl2
ADP-Glo™ Kinase Assay and EGFR Kinase Enzyme System (V9261)
  • 1 x 10μg EGFR Kinase Enzyme System (V3831) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)

Recombinant human EGFR (amino acids 695-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. EGFR is the receptor for members of the EGF family and is a transmembrane glycoprotein that has tyrosine kinase activity.

  • Easily Screen and Profile EGFR Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Molecular Weight (g/mol) 89
Content And Storage <-65°C
Quantity 10 μg
Form Fluid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.