Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Exonuclease III

Catalog No. PRM1811 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25,000 U
5000 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-M1811 5000 U
PR-M1815 25,000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR-M1811 Supplier Promega Supplier No. M1811
Add to Cart
Add to Cart

Includes

  • Part No. M1811: Exonuclease III 10X Buffer (1 x 1mL); Exonuclease III (5000U).
  • Part No. M1815: Exonuclease III 10X Buffer (5 x 1mL); Exonuclease III (25,000U).

Exonuclease III is a 3' → 5' exonuclease specific for dsDNA.

  • A 3′→5′ Exonuclease Specific for Double-Stranded DNA
  • Control deletion rate by varying incubation temperature
  • Inactivate Exonuclease III by heating at 75°C for 10 minutes
  • Supplied at a concentration of 150-200u/μL

Specifications

Concentration 150 to 200 U/μL
Color Colorless
Components 10X Reaction Buffer: 660mM Tris HCl (pH 8 at 25°C), 6.6mM MgCl2
Purity >90%
pH 7.5
For Use With (Application) Generating nested sets of deletions with double-stranded, linear DNA, preparation of single-stranded DNA
Storage Buffer 20mM Tris HCl (pH 7.5 at 25°C), 1mM DTT, 100mM KCl, 50% Glycerol
Content And Storage -30°C to -10°C
Source Recombinant E. coli strain
Quantity 5000 U
Product Type Exonuclease III
Molecular Weight (g/mol) 28000
Inhibitors 50mM NaCl
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.