Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Extra Columns for Selected Wizard™ Plus Systems

Catalog No. p-128028 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
100/Ea.
250/Ea.
50/Ea.
Diameter (English):
0.5 in.
0.75 in.
1.25 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Diameter (English)
PR-A7211 250/Ea. 0.5 in.
PR-A7421 50/Ea. 1.25 in.
PR-A7651 100/Ea. 0.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR-A7211 Supplier Promega Supplier No. A7211
Only null left
Add to Cart
Add to Cart

Eliminates many of the problems associated with standard midi-, mini- or maxi-prep procedures and provides a simple and reliable method for rapid isolation of plasmid DNA.

  • Wizard™ Columns are used with the Wizard™ PCR Preps and Wizard™ Plus DNA Purification Systems
  • Each system contains all necessary reagents and columns for DNA isolations from bacterial culture (using EndA– strains)
  • No organic extractions or ethanol precipitations
  • The purified plasmid can be used directly for automated fluorescent DNA sequencing and restriction digestion without further manipulation

Specifications

Content And Storage 15°C to 30°C
Diameter (English) 0.5 in.
Diameter (Metric) 1.3cm
For Use With (Equipment) Wizard™ Plus system
Product Type Column
Quantity 250/Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.