Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega FAK Kinase Enzyme System

Catalog No. p-4672074 Shop All Promega Corporation Products
Click to view available options
Quantity:
1/Ea.
10 μg

Includes

  • 1 x 10μg CSK Kinase Enzyme System (V1971) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)

Recombinant human FAK was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. FAK is involved in signal transduction from integrin-enriched focal adhesion sites that mediate cell contact with the extracellular matrix.

  • Easily Screen and Profile FAK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Molecular Weight (g/mol) 35
Content And Storage <-65°C
Quantity 10 μg
Form Fluid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.