Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega FastBreak™ Cell Lysis Reagent

Catalog No. p-2652323 Shop All Promega Corporation Products
Click to view available options
Quantity:
15 mL
60 mL
100 mL

FastBreak Cell Lysis Reagent allows efficient, gentle lysis of E. coli cultures without the need for centrifugation or mechanical disruption.

  • Quick E. coli Cell Culture Lysis, No Centrifugation or Mechanical Disruption Required
  • Eliminate centrifugation and mechanical disruption
  • Simple, add-and-incubate protocol
  • Compatible with manual or robotic platforms

  • Patent Pending.
  • This product is licensed for use under U.S. Patent 6,174,704.
  • Certain applications of this product are covered by patents issued and applicable in certain countries. Because purchase of this product does not include a license to perform any patented application, users of this product may be required to obtain a patent license depending upon the particular application and country in which the product is used.

Specifications

Concentration 10X
Product Type Cell Lysis Reagent
Content And Storage 4°C to 25°C
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.