Learn More
Description
- Reagent Used with Flexi(R) Cloning System
- Required to screen Flexi(R) Vector clones for presence of protein-coding region of interest
- Also can be used to cleave PCR products prior to cloning into Flexi(R) Vectors or transfer protein-coding regions between Flexi(R) Vectors
Specifications
Specifications
| Content And Storage | -30°C to -10°C |
| For Use With (Equipment) | Flexi cloning system |
| Includes | 10X Flexi Enzyme blend (SgfI and Pmel)1 x 100μL, Nuclease-free Water 2 x 1,250μL, 5X Flexi Digest Buffer 1 x 1mL |
| pH | 7.5 |
| Product Type | Enzyme Blend |
| Quantity | 25 μL |
| Form | Fluid |
Licensed under U.S. Patent 5,945,288.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.