Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega GeneClip™ U1 Hairpin Cloning Systems

Catalog No. PRC8760 Shop All Promega Corporation Products
Change view
Click to view available options
Vector:
Hygromycin
Puromycin
hMGFP
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Vector
PRC8760 Puromycin
PRC8790 hMGFP
PRC8770 Hygromycin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRC8760 Supplier Promega Supplier No. C8760

Includes

2X Rapid Ligation Buffer, 200μL; Nuclease-free Water, 1.25mL; Oligo Annealing Buffer, 1mL; pGeneClip Vector (varies with system), 1.2μg; T4 DNA Ligase, 100U

Designed for fast and easy cloning of human hairpin target sequences to allow expression of siRNA target sequences in human cells

  • Use pGeneClip™ vectors containing the human U1 promotor, which allows transcription of the hairpin target sequences and generation of hairpin siRNA in vivo.
  • Utilize predigested vectors to eliminate time-consuming vector preparation
  • Allow easier identification of desired clones using a <i>Pst</i> I digestion
  • Available with a choice of vectors
  • The hMGFP System enables easy determination of transfection efficiency
  • The other systems offer antibiotic marker options for selecting stably transfected eukaryotic cells so that experimental results do not depend on transfection efficiency
  • Kit Contents: 200&mu;L 2X Rapid Ligation Buffer, 1.25mL Nuclease-free water, 1mL Oligo Annealing Buffer, 1.2&mu;g pGeneClip Puromycin Vector, 100U T4 DNA ligase

  • For nonprofit and noncommercial research use only. With respect to commercial research use or use by for-profit organizations, or any diagnostic or therapeutic uses, please contact Promega Corporation for supply and licensing information.
  • Licensed under U.S. Patent 6,573,099 and its foreign equivalents from Benitec Australia Ltd and CSIRO.
  • This product is covered under license from Carnegie Institution of Washington under U.S. Pat. Nos. 6,506,559, 7,538,095, 7,560,438; Australian Patent 743798 and other patents pending. Commercial use of this product may require a separate license from Carnegie.
  • For research use only. With respect to any and all other uses, please contact Promega Corporation for supply and licensing information.
  • Certain applications of this product may require licenses from others.

Specifications

Content And Storage Store at −20°C.
Description U1 Hairpin Cloning System—Puromycin
For Use With (Application) Fast and easy cloning of Human hairpin target sequences to allow expression of siRNA target sequences in Human cells
Includes 2X Rapid Ligation Buffer, 200μL; Nuclease-free Water, 1.25mL; Oligo Annealing Buffer, 1mL; pGeneClip Vector (varies with system), 1.2μg; T4 DNA Ligase, 100U
No. of Reactions 20
No. of Samples 20
pH 7.4
Promoter U1
Vector Puromycin
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.