Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Glycosidases
Shop All Promega Corporation Products
Click to view available options
Quantity:
10,000 U
50,000 U
Description
- Characterize Protein Glycosylation
- Monitor protein trafficking
- Characterize glycan location and structure
- Does not cleave complex glycans
- Supplied at a concentration of 500u/μL
Specifications
Specifications
| Concentration | 500 U/μL |
| For Use With (Application) | Determining the state of Protein Glycosylation, characterizing Glycan structure |
| Formulation | 1 x 10000U Endo H, 1 x 1mL 10X Endo H Reaction Buffer, 1 x 1mL 10X Denaturing Solution |
| Content And Storage | -30°C to -10°C |
| Quantity | 10,000 U |
| Product Type | Endoglycosidase H |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction