Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega GoTaq™ Probe qPCR Master Mix

Catalog No. PRA6101 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
1000 Reactions
200 Reactions
Quantity:
2 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions Quantity
PRA6101 200 Reactions 2 mL
PRA6102 1000 Reactions 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRA6101 Supplier Promega Supplier No. A6101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GoTaq™ Probe qPCR Master Mix is a ready-to-use stabilized 2X formulation containing GoTaq™ Hot Start Polymerase, MgCl2, dNTPs and proprietary reaction buffer.

  • Convenient qPCR Amplification Using Hydrolysis Detection Probes
  • Ready-to-use mix of optimized components for robust amplification
  • Compatible with both fast and standard cycling methods
  • Sensitive detection for quantitation over a broad dynamic range

Specifications

Concentration 2X
Content And Storage • 2X GoTaq Probe qPCR Master Mix (dTTP), 2 x 1 mL
• 30μM CXR Reference Dye, 100 μL
• Nuclease-free Water, 2 x 1.25 mL

Store at –30° to –10°C.
Detection Method Primer-probe
Form Frozen
Format Liquid
PCR Method qPCR
Reaction Speed Fast or Standard
Technique qPCR
For Use With (Equipment) Real-Time PCR Systems
Kit Type QPCR Detection Kit
Optimal Reaction Temperature 60°C
Passive Reference Dye CXR Reference Dye (Separate Tube)
Quantity 2 mL
Shipping Condition Frozen
For Use With (Application) Gene Expression
Thermostability GoTaq DNA Polymerase activation: 95°C for 2 minutes, Denaturation: 95°C for 15 seconds (standard cycling) or 3 seconds (fast cycling), Annealing and Extension: 60°C
Compatibility Both standard and fast instrument cycling programs
Hot Start Antibody-mediated Hot-start Chemistry
No. of Reactions 200 Reactions
Sample Type DNA Template
Inhibitor Tolerance (Simple) High
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.