Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega HaloCHIP™ Protein-DNA Covalent Capture System
Shop All Promega Corporation Products
Click to view available options
:
20 reactions
Description
- Covalent Capture of Protein:DNA Complexes, No Antibodies Needed
- No need to make or purchase qualified antibodies
- Results in 24-48 hours with fewer steps, minimizing potential errors
Specifications
Specifications
Content And Storage | COMPONENTS AT DIFFERENT TEMPS |
No. of Reactions | 20 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction