Learn More
Description
- Detects HDAC Class I and II Enzyme Activity
- Simple add-mix-measure protocol
- Use purified enzymes, extracts or cells directly in culture plates
- Screening System includes HeLa Nuclear Extract
Specifications
Specifications
| Quantity | 50 mL |
| pH | 8 |
| Product Type | Assay |
| Content And Storage | 1 x 125mL HDAC-Glo™ I or II Buffer, 5 x 1 Vial HDAC-Glo™ I or II substrate, 1 x 50μL developer reagent, 1 x 10μL Trichostatin A. Store at -30°C to -10°C |
| For Use With (Application) | Determining HDAC inhibitor potency using purified enzymes, extracts or cells directly in culture plates |
| Form | Fluid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.