Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ImProm-II™ Reverse Transcription

Catalog No. p-127565 Shop All Promega Corporation Products
Click to view available options
Product Type:
ImProm-II Reverse Transcriptase
ImProm-II Reverse Transcription System
No. of Reactions:
10
100
500
This item is not returnable. View return policy

Catalog No. PRA3800

Supplier: Promega A3800

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Formulated for stand-alone applications or for easy transition to gene-specific target amplification.

  • Reverse transcribes RNA templates from total RNA, poly(A)+ mRNA, or synthetic transcript RNA
  • Enables robust, full-length cDNA synthesis for the analysis of rare or long messages (up to 8.9kb)
  • Uses volumes of 1–20μL from the initial RT reaction in coupled or uncoupled PCR reactions
  • May also be used for the incorporation of regular, Cy™3-modified, Cy5-modified, and amino-allyl-modified nucleotides for microarray applications
  • Refill components available separately

Specifications

Content And Storage -30°C to -10°C
For Use With (Application) Reverse transcription of RNA templates from total RNA, poly(A)+ mRNA, or synthetic transcript RNA
Includes Upstream control primer 100μL, Downstream control primer 100μL, MgCl2 1.2mL, Oligo(dT)15 Primer 50μg, dNTP mix 320μL, Random primers 50μg, 1.2kb kanamycin positive control RNA 5μg, ImProm-II 5X reaction buffer 600μL, ImProm-II reverse transcriptase 100μL, Recombinant RNasin ribonuclease inhibitor 2500U, Nuclease-free water 2 × 1250μL
No. of Reactions 100
pH 7
Product Type ImProm-II Reverse Transcription System
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.