Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega LYN B Kinase Enzyme System

Catalog No. p-4685441 Shop All Promega Corporation Products
Click to view available options
Quantity:
10 μg
1000 Assays

Recombinant full-length human LYN B was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. LYN is a 56kDa tyrosine kinase that is similar to mouse T-lymphocyte-specific tyrosine kinase p56lck, v-yes, v-fgr and v-src.

  • Easily Screen and Profile LYN B Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Molecular Weight (g/mol) 85
Content And Storage -70°C
Quantity 10 μg
Form Fluid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.