Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega M-MLV Reverse Transcriptase, RNase H Minus, Point Mutant
Point mutation of M-MLV RT resulting in no RNase H activity. Suitable for rare or long transcripts.
$179.97 - $2383.25
Specifications
Content And Storage | −30 to −10°C |
---|---|
Description | M-MLV Reverse Transcriptase |
Form | Liquid |
For Use With (Application) | cDNA Synthesis |
Source | Recombinant E. coli Strain |
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity & Availability | ||||
PRM3682
|
Promega
M3682 |
M-MLV Reverse Transcriptase | 10000U |
Each for $428.32
|
|
||||
PRM3681
|
Promega
M3681 |
M-MLV Reverse Transcriptase | 2500U |
Each for $179.97
|
|
||||
PRM3683
|
Promega
M3683 |
M-MLV Reverse Transcriptase | 50000U |
Each for $2,383.25
|
|
||||
Description
- High Yields of cDNA from Long RNA Templates (>5kb)
Specifications
−30 to −10°C | |
Liquid | |
Recombinant E. coli Strain |
M-MLV Reverse Transcriptase | |
cDNA Synthesis | |
Tris-HCl (pH 7.5 at 25°C) 20mM, NaCl 200mM, EDTA 0.1mM, DTT 1mM, 0.01% Nonidet™ P40™ and Glycerol 50% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title