Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Maxwell™ 16 LEV simplyRNA Blood Kit

Catalog No. pras1310 Shop All Promega Corporation Products
Click to view available options
Quantity:
48 Preps

Includes

1-Thioglycerol 900μL, Cell lysis solution 4 x 100mL, Maxwell 16 LEV Cartridges (48), LEV Plungers (50), 0.5mL Elution Tubes (50), Blue Dye 50μL, Proteinase K (PK) solution 2 x 1mL, Lysis Buffer 20mL, Nuclease-free water 25mL, Homogenization solution 30mL and DNase I (lyophilized) 2 vials

The Maxwell 16 LEV simplyRNA Kits are for use with the Maxwell 16 Instrument configured with the LEV High Strength Magnetic Rod and Plunger Bar Adaptor for automated RNA purification after minimal lysate handling.

  • Automated RNA Purification from 1-16 Blood, Cell or Tissue Samples
  • Simple preprocessing; no centrifugation
  • Used with the Maxwell(R) 16 Instrument, automated protocol generates consistent results and sufficient yield for downstream assays
  • Use with fresh or frozen cells or tissues

Specifications

Content And Storage 15°C to 30°C
For Use With (Application) Isolation of total RNA from fresh whole Blood collected in EDTA tubes
Includes 900μL 1-Thioglycerol, 4 x 100mL Cell Lysis solution, 48 Maxwell™ 16 LEV cartridges, 50 LEV plungers, 0.5mL 50 elution tubes, 50μL blue dye, 2 x 1mL Proteinase K solution, 20mL Lysis Buffer, 25mL Nuclea
Quantity 48 Preps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.