Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Maxwell™ 16 LEV SimplyRNA Purification Kits
Shop All Promega Corporation Products
Click to view available options
Quantity:
48 Preps
Product Type:
LEV simplyRNA Tissue Kit
Description
- Automated RNA Purification from 1-16 Blood, Cell or Tissue Samples
- Simple preprocessing; no centrifugation
- Used with the Maxwell(R) 16 Instrument, automated protocol generates consistent results and sufficient yield for downstream assays
- Use with fresh or frozen cells or tissues
Certifications
FDA 510K Cleared for medical use
Specifications
Specifications
Content And Storage | 15°C to 30°C |
For Use With (Application) | Gene expression, used in many areas of research including translation, quantitative RT-PCR |
Includes | Thioglycerol 1 x 900μL, Maxwell™16 LEV cartridge 48 x 1, LEV plungers 1 x 50, elution tubes 1 x 50, blue dye 1 x 50μL, Lysis Buffer 1 x 20mL, Nuclease-free Water 1 x 25mL, homogenization solution 1 x 3 |
Product Type | LEV simplyRNA Tissue Kit |
Quantity | 48 Preps |
For Research Use Only (RUO)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction