Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega NAD/NADH-Glo™ Assay

Catalog No. PRG9072 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG9072 50 mL
PRG9071 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRG9072 Supplier Promega Supplier No. G9072
Add to Cart
Add to Cart

The NAD/NADH-Glo™ Assay is a bioluminescent, homogeneous single-reagent-addition assay for detecting total oxidized and reduced nicotinamide adenine dinucleotides and determining their ratio in biological samples or in defined enzyme reactions.

  • Bioluminescent Assays to Detect NAD(P)+ and NAD(P)H Levels in Cells
  • Homogeneous, single-reagent-addition assays
  • Easily adapted for high throughput screening formats

Specifications

Quantity 50 mL
Content And Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.