Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega pCMVTNT™ Vector
Shop All Promega Corporation Products
Click to view available options
Quantity:
20 μg
Description
- Convenient Expression of Cloned Genes In Vitro or In Vivo
- Express cloned genes in vitro or in vivo from SP6- or T7-based coupled in vitro transcription/translation systems
Specifications
Specifications
| Promoter | CMV |
| Content And Storage | Below -10°C |
| Quantity | 20 μg |
| Vector | pCMVTnT vector |
| For Use With (Application) | Convenient expression of cloned genes using both in vivo and in-vitro expression systems |
Licensed for research and laboratory use only under U.S. Patent 5,547,862.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction