Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega pGL3 Luciferase Reporter Vectors

Catalog No. PRE1741 Shop All Promega Corporation Products
Change view
Click to view available options
Vector:
pGL3-control vector
pGL3-promoter vector
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Vector
PR-E1741 pGL3-control vector
PR-E1761 pGL3-promoter vector
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR-E1741 Supplier Promega Supplier No. E1741

Includes

Vector DNA, 20μg; Bacterial strain JM109, 500μL

Four firefly luciferase reporter vectors.

  • Four Reporter Vectors That Contain luc+, a Modified Firefly Luciferase Gene
  • Unique NcoI site located at 5′ end of luc+ gene will create fusions with reporter gene
  • SmaI site in the MCS means blunt-ended inserts can be ligated into the MCS and cut on either side by other restriction endonucleases
  • XbaI site just downstream of luc+ gene facilitates insertions into the 3′ untranslated region of mRNA or subcloning of the luciferase gene

  1. U.S. Patent 5,670,356.
  2. The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.

Specifications

Promoter SV40
Reporter Gene Luciferase
Content And Storage -30°C to -10°C
Quantity 20 μg
Vector pGL3-control vector
For Use With (Application) Quantitative analysis of factors that potentially regulate mammalian gene expression
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.