Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega pGL3 Luciferase Reporter Vectors
Shop All Promega Corporation Products
Click to view available options
Vector:
pGL3-basic vector
pGL3-control vector
pGL3-enhancer vector
pGL3-promoter vector
Description
Includes
Vector DNA, 20μg; Bacterial strain JM109, 500μL
- Four Reporter Vectors That Contain luc+, a Modified Firefly Luciferase Gene
- Unique NcoI site located at 5′ end of luc+ gene will create fusions with reporter gene
- SmaI site in the MCS means blunt-ended inserts can be ligated into the MCS and cut on either side by other restriction endonucleases
- XbaI site just downstream of luc+ gene facilitates insertions into the 3′ untranslated region of mRNA or subcloning of the luciferase gene
- U.S. Patent 5,670,356.
- The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.
Specifications
Specifications
Promoter | SV40 |
Reporter Gene | Luciferase |
Content And Storage | -30°C to -10°C |
Quantity | 20 μg |
Vector | pGL3-enhancer vector |
For Use With (Application) | Quantitative analysis of factors that potentially regulate mammalian gene expression |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction