Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega pGloSensor™-10F Linear Vector
Shop All Promega Corporation Products
Click to view available options
Quantity:
1 μg
Description
- Detect and Measure Protease Activities
- Luminescent assay
- Quickly create your own protease substrates
- Easy quantitation, high sensitivity and wide dynamic range
Specifications
Specifications
For Use With (Equipment) | Protease-Glo™ assay |
Restriction Site | SgfI and PmeI |
Content And Storage | -30°C to -10°C |
Quantity | 1 μg |
Vector | pGloSensor-10F linear vector |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction