Learn More
Promega PKCβ II Kinase Enzyme System
Catalog No. p-4685413 Shop All Promega Corporation Products
Click to view available options
Quantity:
1/Ea.
10 μg
Catalog No. PRV3741
Supplier: Promega V3741
Only null left
Add to Cart
Add to Cart
Recombinant full-length human PKC beta II was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKC beta II is a member of the protein kinase C (PKC) family of serine- and threonine-specific protein kinases.
  • Easily Screen and Profile PKCβII Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Content And Storage <-65°C
For Use With (Application) Involved in many different cellular functions such as B cell activation, apoptosis induction, endothelial cell proliferation and intestinal sugar absorption
Quantity 10 μg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.