Learn More
Promega PKCζ Kinase Enzyme System
Catalog No. p-4672124 Shop All Promega Corporation Products
Click to view available options
Quantity:
1/Ea.
10 μg
Includes
PKCζ Kinase Assay:
  • Kinase: Recombinant Human PKCζ, 10μg (full length), MW: ≅93kDa
  • Substrate: CREBtide (KRREILSRRPSYR) derived from human CREB1 isoform A (amino acid 109 to 121)
  • Other: Reaction Buffer, DTT
ADP-Glo Kinase Assay: Reagent, buffer, substrate, Ultra Pure ATP (10mM), ADP (10mM)
Recombinant full-length human PKC zeta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKC zeta is an atypical isoform of the PKC family.
  • Easily Screen and Profile PKCζ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Content And Storage <-65°C
Quantity 10 μg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.