Learn More
Description
Includes
DNA Polymerase I Large (Klenow) Fragment; Prime-a-Gene dATP; Prime-a-Gene dCTP; Prime-a-Gene dGTP; Prime-a-Gene dTTP; Labeling 5X Buffer; BSA, nuclease-free; Prime-a-Gene control DNA; Nuclease-free water
- Complete Kit for Random-Primed Labeling of Linear Template DNA, Excluding Radionucleotides
- Ready-to-use reagents for random-primed labeling of linear DNA, including random hexamer primers (excludes radionucleotides)
- Probes generated with high specific activities >1 x 109cpm/μg
Specifications
Specifications
| Content And Storage | -30°C to -10°C |
| Includes | DNA Polymerase I large (Klenow) fragment, prime-a-gene dATP, prime-a-gene dCTP, prime-a-gene dGTP, prime-a-gene dTTP, labeling 5X Buffer, BSA, Nuclease-free, prime-a-gene control DNA, Nuclease-free Water |
| No. of Reactions | 30 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.