Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Protein Deglycosylation Mix
Click to view available options
Quantity:
20 Reactions
Description
Mixture of five (5) protein deglycosidases: PNGase F, O-Glycosidase, Neurominidase, β1-4 Galactosidase, β-N-Acetylglucosaminidase). Provides suffucient deglycosidase for 20 reactions.
Specifications
Specifications
Components | 100μL Protein Deglycosylation mix (PNGase F, O-Glycosidase, Neuraminidase, beta1-4 Galactosidase, beta-N-Acetylglucosaminidase), 1mL 10X Denaturing solution, 1mL 10X Deglycosylation Reaction Buffer (0.5M Sodium Phosphate (1X pH 7.5 at 25 |
For Use With (Application) | Removing Glycans from both O-linked and N-linked glycosylation sites |
Storage Buffer | 20mM Tris HCl (pH 7.5 at 25°C), 50mM NaCl, 5mM EDTA |
Content And Storage | 2°C to 10°C |
Quantity | 20 Reactions |
Product Type | Protein Deglycosylation Mix |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction