Learn More
Promega Protein Kinase C Activator
Catalog No. PRV1171
Click to view available options
Quantity:
5 mg
Phorbol 12-myristate 13-acetate (PMA) is a specific activator of group A (alpha, betaI, betaII, gamma) and group B (delta, epsilon, eta, theta) Protein Kinase Cs (PKCs).
  • Phorbol 12-Myristate 13-Acetate
  • Specific activator of group A (α, βI, βII, γ) and group B (δ, ε, η, θ) Protein Kinase Cs
  • Supplied as a dried material
Specifications
Quantity 5 mg
Content And Storage -30°C to -10°C
Kit Contents 1 x 5mg PMA
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.