Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ReliaPrep™ Blood gDNA Miniprep System

Catalog No. p-4390061 Shop All Promega Corporation Products
Change view
Click to view available options
No. of Reactions:
100 Preps
250 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
PRA5082 250 Preps
PRA5081 100 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRA5082 Supplier Promega Supplier No. A5082
Only null left
Add to Cart
Add to Cart

Includes

ReliaPrep Binding Columns, Collection Tubes, Cell Lysis Buffer (CLD), Proteinase K (PK) Solution, Binding Buffer (BBA), Column Wash Solution (CWD), Nuclease-Free Water

The ReliaPrep Blood gDNA Miniprep System provides a complete, ready-to-use method for purification of gDNA from up to 200ul of blood or body fluid, consistently isolating pure, intact gDNA without the use of alcohol washes or precipitations.

  • Purifies DNA from up to 200μL Blood or Body Fluids
  • No ethanol, eliminating alcohol inhibition and carryover
  • Good recovery and purity in as little as 50μL elution
  • Consistent results from run-to-run and user-to-user

Specifications

Content And Storage 15°C to 30°C
For Use With (Application) Purifies genomic DNA from up to 200μL of Blood or body fluid, consistently isolating pure, intact genomic DNA without the use of Alcohol washes or precipitations
Includes 5 Pack ReliaPrep™ binding columns, 5 pack collection tubes, 55mL Cell Lysis Buffer, 5.5mL Proteinase K solution, 68.75mL Binding Buffer, 412.5mL Column wash solution, 50mL Nuclease-free Water
No. of Reactions 250 Preps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.