Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Alcohol Wash

Catalog No. p-3336513 Shop All Promega Corporation Products
Click to view available options
Quantity:
25 mL
160 mL
Product Type:
Lysis Buffer, Blood
Paramag Particles, Fixed Yield
Paramagnetic Particles

Catalog No. PRMD1392

Supplier: Promega MD1392

Only null left
Add to Cart
Add to Cart

Alcohol Wash for MagneSil Blood Genomic, Max Yield System (Cat.# MD1360), and MagneSil ONE, Fixed Yield Blood Genomic System (Cat.# MD1370).

  • Lysis Buffer Used With Automated Blood DNA Purification Systems
  • Component of the MagneSil Blood Genomic, Max Yield System (Cat.# MD1360), and MagneSil ONE, Fixed Yield Blood Genomic System (Cat.# MD1370).

†Also supplied with Mfr. No. A4080.

Specifications

Content And Storage 15°C to 30°C
For Use With (Equipment) Magnesil™ Blood Genomic system, Magnesil™ one system
For Use With (Application) Automated high-throughput DNA purification from Anti-coagulated whole Blood
Product Type Lysis Buffer, Blood
Quantity 160 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.