Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Restriction Enzymes - Alu I (B)

Catalog No. PRR6281
Click to view available options
Quantity:
500 U

Capable of digesting DNA in 15 minutes or less. (AG/CT TC\GA)

  • Capable of digesting DNA in 15 minutes or less

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Specifications

Concentration 10 U/μL
Components 1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 500U AluI
Incubator Temperature 37°C
pH 7.4
For Use With (Application) Capable of digesting DNA in <15 min.
Storage Buffer 10mM Tris HCl (pH 7.4), 100mM Sodium Citrate, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Content And Storage -30°C to -10°C
Quantity 500 U
Cut Site AG.CT TC.GA
Product Type Alul
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.