Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Restriction Enzymes - Pst I Blue/White Cloning Qualified (H)
Click to view available options
Quantity:
15,000 U
3000 U
Concentration:
10 U/μL
Description
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
- Blue/white cloning qualified, provides a higher level of quality control for enzymes used in cloning applications
‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
Specifications
Concentration | 10 U/μL |
Components | 1 x 1mL 10X Buffer H, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 3000U PstI, 1 x 0.25mL Multi-Core™ 10X Buffer |
Incubator Temperature | 37°C |
pH | 7.3 |
For Use With (Application) | Provides a higher level of quality control for Enzymes used in cloning applications |
Storage Buffer | 10mM Tris HCl (pH 7.3), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.15% Triton™ X-100, 0.5mg/mL BSA, 50% Glycerol |
Content And Storage | -30°C to -10°C |
Quantity | 3000 U |
Cut Site | CTGCA.G G.ACGTC |
Product Type | PstI |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction