Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega ROCK1 Kinase Enzyme System
Shop All Promega Corporation Products
Click to view available options
Quantity:
1/Ea.
10 μg
Description
- Easily Screen and Profile ROCK1 Kinase Inhibitors
- Includes kinase, substrate and reaction buffer
- Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
| Molecular Weight (g/mol) | 85 |
| Content And Storage | <-65°C |
| For Use With (Application) | Diverse cellular functions, including smooth muscle contraction, actin cytoskeleton organization, cell adhesion and motility and gene expression |
| Quantity | 10 μg |
| Form | Fluid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction