Learn More
Promega RON Kinase Enzyme System
Catalog No. p-4672104 Shop All Promega Corporation Products
Click to view available options
Quantity:
1/Ea.
10 μg
Includes
  • Kinase: Recombinant Human RON, 10μg (amino acids 983-end), MW: ∽71kDa
  • Substrate: Axltide (KKSRGDYMTMQIG) derived from mouse Insulin receptor substrate 1 (amino acid 979 to 989)
  • Other: Reaction Buffer, DTT
Recombinant human RON (amino acids 983-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. RON is a macrophage stimulating 1 receptor c-met-related tyrosine kinase.
  • Easily Screen and Profile RON Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Content And Storage <-65°C
Quantity 10 μg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.