Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega StemElite™ Gene Expression System

A real-time QPCR system for the detection and relative quantification of RNA expression levels associated with the differentiation state or potency of cells

Supplier:  Promega B1001

Catalog No. PRB1001


Includes: StemElite 2X Master Mix, 1mL; Nuclease-free water, 1.25mL

Description

Description

  • Optimized to quantitatively amplify a two-color duplex, enabling the user to amplify a transcript of interest as well as a reference gene in a single reaction
  • Achieves sensitive real-time detection and relative quantification of RNA expression levels for monitoring of stem cell differentiation in a multiplexed amplification
  • Quantitatively amplifies a two-color duplex
  • Amplifies in a single tube the transcript of interest and reference transcript (GAPDH and Actb)
  • Reduces the number of reactions required for the measurement of multiple transcripts
  • Improves experimental data quality by measuring all the transcripts in the same well

Specifications

Specifications

StemElite™ 2X master mix (1 x 1mL), Nuclease free Water (1 x 1250μL). Store at -20°C
RT-qPCR reaction kit
100
Gene Expression
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

  • U.S. Patent 6,242,235, Australian Patent 761757, Canadian Patent 2,335,153 and other patents and patents pending.
  • Licensed under U.S. Patents 5,338,671 and 5,587,287 and corresponding patents in other countries. For Research Use Only. Not for use in diagnostic procedures.
  • The purchase of this product conveys to the buyer the limited, nonexclusive, nontransferable right (without the right to resell, repackage, or further sublicense) under U.S. Published Patent Appln. 20020150900 and U.S. Patents 5,432,272; 6,617,106 and 6,140,496 to use the product. No other license is granted to the buyer whether expressly, by implication, by estoppel or otherwise. In particular, the purchase of this product does not include or carry any right or license to sell this product. For information on purchasing a license for other uses, please contact Promega Corporation, Business Development, 2800 Woods Hollow Road, Madison, WI 53711, or EraGen Biosciences, Corporate Licensing, 918 Deming Way, Suite 201, Madison, WI 53717. Phone (608) 662-9000; Fax (608) 662-9003.