Learn More
Description
- Adds Mononucleotides to the Terminal 3′-OH of ssDNA and Releases PPi
- Adds mononucleotide tails to any type of 3′ end (3′ and 5′ overhangs or blunt ends) due to the presence of 1mM CoCl2 in the reaction buffer
- Quality tested for labeling of apoptotic DNA ends using the procedure outlined in the DeadEnd™ Fluorometric TUNEL System Technical Bulletin #TB235 for each lot of enzyme
- Supplied at a concentration of 30u/μL
Specifications
Specifications
| Concentration | 30 U/μL |
| Color | Colorless |
| Components | 5X Reaction Buffer: 500mM Cacodylate Buffer (pH 6.8 at 25°C), 5mM CoCl2, 0.5mM DTT |
| pH | 6.4 |
| For Use With (Application) | Tailing reactions to add complementary Homopolymer tails to DNA vectors and cDNA, 3' end-labeling, TUNEL Assays |
| Storage Buffer | 50mM Potassium Phosphate (pH 6.4), 100mM NaCl, 1mM beta-mercaptoethanol, 0.1% Tween™ 20, 50% Glycerol |
| Content And Storage | -30°C to -10°C |
| Source | Recombinant E. coli strain |
| Quantity | 300 U |
| Product Type | Terminal Deoxynucleotidyl Transferase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.