Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega ViviRen™ In Vivo Renilla Luciferase Substrate
Shop All Promega Corporation Products
Click to view available options
Substrate Quantity:
0.37mg
3.7mg
Description
- Engineered Coelenterazine-Based Substrate for Animal Imaging
- High-quality substrate eliminates potential interference in assays due to the presence of endotoxins
- Available in multiple sizes to accommodate a variety of experimental settings
Specifications
Specifications
For Use With (Application) | In-vivo bioluminescence imaging applications |
Quantity | 0.37mg |
Substrate Type | Renilla Luciferase Substrate |
Storage Requirements | −30 to −10°C |
Substrate Quantity | 0.37mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction