Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Wizard™ Lambda Preps DNA Purification System1

Catalog No. PRA7290 Shop All Promega Corporation Products
Change view
Click to view available options
:
20 Mini Columns
Catalog No.
PR-A7290 20 Mini Columns
1 options

Catalog No. PR-A7290

Supplier: Promega A7290

Includes

  • Nuclease Mixture, 500μg
  • Nuclease Resuspension Solution, 1mL
  • Phage Precipitant, 80mL
  • Phage Buffer, 10mL
  • Wizard Minicolumns, 20
  • Wizard Lambda DNA Purification Resin, 20mL
  • 3cc syringe barrels, 25

For fast, reliable purification of λ DNA from plate or liquid culture lysate in 1 1/2 hours or less

  • Produces high-purity λ DNA that may be used in restriction digestions, ligations, and sequencing without any further purification
  • Contains sufficient reagents and columns for 20 isolations from 10mL of lysate
  • Can be used to process multiple samples at one time when used with a vacuum manifold such as the Vac-Man™ (PR-A7231) or Vac-Man Jr. (PR-A7660) Laboratory Vacuum Manifold
  • Store at 22° to 25°C

1U.S. Patents 5,658,548 and 5,808,041; Australian Patents 689815 and other patents and patents pending.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.