Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Wizard™ MagneSil™ Plasmid Purification System
Shop All Promega Corporation Products
Click to view available options
Quantity:
100 mL
100 x 96 Preps
4 x 96 Preps
8 x 96 Preps
Description
Includes
- MagneSil BLUE, MagneSil RED, plus necessary solutions, buffers, and collection plates for plasmid purification
- MagneSil RED (also available separately)
Requires
- Orbital microplate shaker
- Magnetic workstation that will accommodate a microplate, such as the MagnaBot™ 96 Magnetic Separation Device (PR-V8151, sold separately)
- Rapid Isolation of Plasmid DNA in 96-Well Format
- Process multiple plates without user intervention
- Validated automated methods available
- U.S. Patents 6,284,470 and 7,078,224; Australian Patent 778486 and other patents pending.
- U.S. Patents 6,027,945 and 6,368,800; Australian Patent 732756 and other patents and patents pending.
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Format | 96-well Plate |
| For Use With (Equipment) | Wizard MagneSil Plasmid Purification System |
| For Use With (Application) | Simple and reliable method for the rapid isolation of plasmid DNA |
| Includes | 50mL Cell resuspension solution, 60mL Cell Lysis solution, 60mL neutralization solution, 19mL Magnesil™ blue, 30mL Magnesil™ red, 50mL Elution Buffer, 16 Collection plates, protocol |
| Product Type | Plasmid Purification |
| Quantity | 4 x 96 Preps |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction