Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Wizard™ MagneSil™ Plasmid Purification System

Catalog No. pra1641 Shop All Promega Corporation Products
Click to view available options
Quantity:
100 mL
100 x 96 Preps
4 x 96 Preps
8 x 96 Preps

Catalog No. PRA1641

Supplier: Promega A1641

Only null left
Add to Cart
Add to Cart

Includes

  • MagneSil BLUE, MagneSil RED, plus necessary solutions, buffers, and collection plates for plasmid purification
  • MagneSil RED (also available separately)

Requires

  • Orbital microplate shaker
  • Magnetic workstation that will accommodate a microplate, such as the MagnaBot™ 96 Magnetic Separation Device (PR-V8151, sold separately)

The Wizard™ MagneSil Plasmid DNA Purification System provides a simple, reliable method for rapid isolation of plasmid DNA in a 96-well, high-throughput format.

  • Rapid Isolation of Plasmid DNA in 96-Well Format
  • Process multiple plates without user intervention
  • Validated automated methods available

  • U.S. Patents 6,284,470 and 7,078,224; Australian Patent 778486 and other patents pending.
  • U.S. Patents 6,027,945 and 6,368,800; Australian Patent 732756 and other patents and patents pending.

Specifications

Content And Storage 15°C to 30°C
Format 96-well Plate
For Use With (Equipment) MagneSil RED
For Use With (Application) Simple and reliable method for the rapid isolation of plasmid DNA
Includes Magnesil™ blue, Magnesil™ red, Plus necessary solutions, Buffers and collection plates for plasmid purification
Product Type Plasmid Purification
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.