Learn More
Promega Wizard SV Gel and PCR Clean-Up System and x-tracta™ Gel Extractor Package
Click to view available options
Quantity:
250 Preparation/100 Extractors
50 Preparation/25 Extractors
Description
- Purifies DNA Fragments from Gels or PCR
- Purifies 100bp to 10kb DNA fragments
- Includes gel extractors for added speed and convenience
Specifications
Specifications
Type | Wizard™ SV Gel and PCR Clean up System and x-tracta™ Gel Extractor Package |
Quantity | 250 Preparation/100 Extractors |
Temperature (Metric) Storage | 22°C to 25°C |