Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ProPac™ Elite WCX HPLC Column
GREENER_CHOICE

Catalog No. 03251450
Change view
Click to view available options
Length (Metric):
50 mm
150 mm
250 mm
Diameter (Metric):
2 mm
4 mm
9 mm
Quantity:
1 Column
3 Columns/1 Lot
3 Columns/3 Lots
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Quantity
03-251-450 250 mm 2 mm 1 Column
03-251-448 50 mm 2 mm 1 Column
03-251-449 150 mm 2 mm 1 Column
03-254-078 50 mm 4 mm 1 Column
03-251-452 150 mm 4 mm 1 Column
03-251-453 250 mm 4 mm 1 Column
03-251-454 150 mm 4 mm 3 Columns/1 Lot
03-251-457 150 mm 4 mm 3 Columns/3 Lots
03-254-079 250 mm 4 mm 3 Columns/1 Lot
03-254-080 250 mm 4 mm 3 Columns/3 Lots
03-254-119 150 mm 9 mm 1 Column
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 03-251-450 Supplier Thermo Scientific™ Supplier No. 303026
Add to Cart
Add to Cart

This column provides consistent separations of therapeutic proteins including monoclonal antibodies, antibody drug conjugates, biosimilars and charge variants.

Discover the unique Thermo Scientific™ ProPac™ Elite WCX weak cation exchange (WCX) liquid chromatography column. Designed for protein characterization including therapeutic proteins, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs) and biosimilar proteins, this column offers high resolution, high efficiency separation of proteins and their charge variants.

The unique column chemistry provides excellent performance under a broad range of pH, temperature and mobile phase compositions with excellent recovery and low carryover.

Product Highlights
  • Superior resolution power for proteins, monoclonal antibodies, and associated charge variants
  • High efficiency with reproducible separa-tions
  • High recovery with low carry-over
  • Wide pH operating range: 2 – 12
  • High temperature stability: up to 60°C
  • High throughput
  • Compatible with CX-1 pH Gradient Buffers
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 2 mm
Flow Rate 0.1 to 0.2 mL/min
Length (Metric) 250 mm
Max. Pressure 4500 psi (310 bar)
Mobile Phase Compatibility Recommended: Goods Buffers and NaCl. Minimum 20 mM salt required. Do not rinse with pure DI H2O
Particle Size 5 μm
pH 2 to 12
Pore Size Nonporous
Quantity 1 Column
Temperature 10°C to 60°C
Stationary Phase Carboxylate
Product Line ProPac
Packing Material Nonporous DVB Particles with Hydrophilic Coating
Column Type Ion Exchange
Phase Ion Exchange
Technique Bio-LC
Green Features FSC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.