Learn More
Thermo Scientific™ ProPac™ IMAC-10 HPLC Columns
Description
Resolve target proteins using a single column in a high-resolution gradient run with Thermo Scientific™ ProPac™ IMAC-10 LC Columns. These analytical and semi-preparative HPLC columns provides high-purity separations of metal-binding proteins and high loading capacity for protein purification applications. The use of small, nonporous resin particles results in narrow, high-efficiency peaks, with minimal band broadening due to dispersion. Use loading columns to charge the ProPac IMAC-10 LC columns but avoid contact of the metal solution with the system.
- Retention control by imidazole concentration or pH gradient
- Stable between pH 2 to 12, compatible with common protein purification reagents (nonionic detergents, reducing agents)
- Effective under native or denaturing HPLC conditions
- Wide range of metal-specific applications, including the separation of His-tagged protein aggregation variants, prion peptide analysis and separation of monoclonal antibodies
- Shipped free from any metal and ready to be charged with the metal of your choice.
- Reusable columns with metal tailored specificity
- Resolve target proteins using a single column in a high resolution gradient run
- High purity separations of metal-binding proteins
Specifications
Specifications
| Column Format | Analytical Column |
| Diameter (Metric) | 4 mm |
| Flow Rate | 1 mL/min |
| For Use With | UltiMate 3000 HPLC System |
| Length (Metric) | 250 mm |
| Max. Pressure | 3000 psi (207 bar) |
| Mobile Phase Compatibility | pH 2 to 12, compatible with most reagents used in IMAC protein purifications, including EtOH, Urea, NaCl, non-ionic detergents, glycerol, acetic acid, guanidine, HCl |
| Particle Size | 10 μm |
| pH | 2 to 12 |
| Pore Size | Nonporous |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.