Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propargyl p-toluenesulfonate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
6165-76-0
C10H10O3S
210.25
MFCD01462194
LMBVCSFXFFROTA-UHFFFAOYSA-N
propargyl p-toluenesulfonate,propargyl tosylate,prop-2-yn-1-yl 4-methylbenzenesulfonate,p-toluenesulfonic acid, 2-propynyl ester,2-propyn-1-ol, 4-methylbenzenesulfonate,ccris 9146,p-toluenesulfonic acid propargyl ester,propargylp-toluenesulfonate,propargyl p-toluene sulfonate,acmc-209mv7
22547
prop-2-ynyl 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OCC#C
6165-76-0
C10H10O3S
210.25
MFCD01462194
LMBVCSFXFFROTA-UHFFFAOYSA-N
propargyl p-toluenesulfonate,propargyl tosylate,prop-2-yn-1-yl 4-methylbenzenesulfonate,p-toluenesulfonic acid, 2-propynyl ester,2-propyn-1-ol, 4-methylbenzenesulfonate,ccris 9146,p-toluenesulfonic acid propargyl ester,propargylp-toluenesulfonate,propargyl p-toluene sulfonate,acmc-209mv7
22547
prop-2-ynyl 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OCC#C
Refractive Index 1.5270 to 1.5290 (20°C, 589nm)
Color Colorless to Brown
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 96% min. (GC)
Linear Formula CH3C6H4SO3CH2C≡CH
Beilstein 11, IV, 263
Formula Weight 210.25
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Propargyl p-toluenesulfonate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning