Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Properdin Antibody (10-24) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10064677 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064677 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064677 Supplier Novus Biologicals Supplier No. NB10064677

Mouse Monoclonal Antibody

Properdin Monoclonal specifically detects Properdin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Properdin
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 10-24
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Gene Accession No. P27918
Gene Alias BFD, Complement factor P, complement factor properdin, PFCcomplement, PFD, PROPERDIN
Gene Symbols CFP
Host Species Mouse
Immunogen Native purified factor P from human serum
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 5199
Test Specificity NB100-64677 recognizes human complement factor P, a 53 kDa glycosylated protein present in blood serum. Factor P binds to microbial surfaces and binds and stabilizes the complement component 3- and complement component 5-convertase enzyme complexes. C3b then acts as an opsonin or initiates the complement pathway again creating a positive feedback loop which acts only on the bacterial cell, leaving the host cells unharmed. Deficiency of factor P makes the host more susceptible to bacterial infections, particularly to Neisseria species and meningococcal infections. NB100-64677 does not inhibit function of complement factor P.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.