Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Properdin Antibody (SIM 295-9.3.4), Novus Biologicals™
SDP

Catalog No. NB119839 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119839 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119839 Supplier Novus Biologicals Supplier No. NBP311214
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Properdin Monoclonal specifically detects Properdin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Frozen, Immunoassay.

Specifications

Antigen Properdin
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Frozen), Immunoassay
Classification Monoclonal
Clone SIM 295-9.3.4
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunohistochemistry-Frozen, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias BFD, Complement factor P, complement factor properdin, PFCcomplement, PFD, PROPERDIN
Host Species Mouse
Immunogen Purified Human Properdin
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5199
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.