Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propiolaldehyde diethyl acetal, 97%

Catalog No. aal1076703
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 10160-87-9 | C7H12O2 | 128.17 g/mol

Propiolaldehyde diethyl acetal is used in preparation of 3-boronoacrolein pinacolate, a heterodyne and (E)-3-(tributylstannyl)-2-propenal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propiolaldehyde diethyl acetal is used in preparation of 3-boronoacrolein pinacolate, a heterodyne and (E)-3-(tributylstannyl)-2-propenal.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Air and moisture sensitive. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10160-87-9
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00009237
InChI Key RGUXEWWHSQGVRZ-UHFFFAOYSA-N
PubChem CID 66285
IUPAC Name 3,3-diethoxyprop-1-yne
SMILES CCOC(OCC)C#C

Specifications

Density 0.894
Boiling Point 138°C to 139°C
Flash Point 32°C (89°F)
Refractive Index 1.412
Quantity 1 g
UN Number UN1993
Beilstein 1701566
Synonym 3,3-diethoxy-1-propyne, propargylaldehyde diethyl acetal, 3,3-diethoxypropyne, propiolaldehyde diethyl acetal, 1-propyne, 3,3-diethoxy, 3,3-diethoxy-propyne, propynal diethyl acetal, 1,1-diethoxyprop-2-yne, 1-propyne,3,3-diethoxy, propargyl aldehyde diethylacetal
Solubility Information Slightly soluble in water.
Formula Weight 128.17
Percent Purity 97%
Chemical Name or Material Propiolaldehyde diethyl acetal
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 233-430-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.