Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propper Seracult™ Plus Single Slide Fecal Occult Blood Test
SDP

Catalog No. 23246930 Shop All Propper Manufacturing Co. Products
Click to view available options
Quantity:
100 Pk.
1000 Pk.
20 Pk.
Type:
Developer
Single Slide

Seracult™ Plus Single Slide Fecal Occult Blood Test is a qualitative aid in the diagnosis of colorectal cancer for use with your most high risk patients. Guaiac Fecal Occult Blood Test to aid in the detection of colorectal cancer. Long sample stability. Fast qualitative results.

  • Seracult™ Plus Fecal Occult Blood Test is a highly sensitive qualitative aid in the diagnosis of colorectal cancer.
  • Designed for routine use in physical examinations in doctors offices, hospitals, clinics, and laboratories or in the privacy and convenience of a patient's home.
  • Seracult™ Plus was designed in mind for patients who are at an increased risk of disease.
  • Seracult™ Plus Single Slides are ideal for testing inpatient or single samples at home.
  • Seracult™ Plus is available in a variety of configurations to meet all of your needs.
  • Results are ready instantly.
  • Seracult guaiac paper allows for enhanced readability for early detection of even the smallest amount of blood in the sample.
  • Mailing kits of in-office slides.
  • 28-day sample stability.
  • CLIA Waived.

Specifications

Clia Complexity Waived
Detectable Analytes Hemoglobin
Sample Type Stool
Type Developer
For Use With (Application) Fecal Occult Blood Detection
Includes 20 x 15 mL Bottles of Developer
Quantity 20 Pk.
Test Time 30 to 60 sec.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.