Learn More
Propper Seracult™ Plus Triple Slide Fecal Occult Blood Test
Shop All Propper Manufacturing Co. Products

Description
- Seracult™ Plus Triple Slides Fecal Occult Blood Test is a highly sensitive qualitative aid in the diagnosis of colorectal cancer.
- Designed for routine use in physical examinations in doctors offices, hospitals, clinics, and laboratories or in the privacy and convenience of a patient's home.
- Seracult™ Plus was designed in mind for patients who are at an increased risk of disease.
- Seracult™ Plus Triple Slides are ideal for multiple sample testing at home.
- Results are ready instantly.
- Seracult paper allows for enhanced readability for early detection of even the smallest amount of blood in the sample.
- Mailing kits of in-office slides.
- 28 day sample stability.
Specifications
Specifications
| Detectable Analytes | Hemoglobin |
| Sample Type | Stool |
| Sensitivity | 0.32 mg/100 mL |
| Type | Developer |
| For Use With (Application) | Fecal Occult Blood Detection |
| Includes | 20 x 15 mL Bottles of Developer |
| Quantity | 20 Pk. |
| Test Time | 60 sec. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.